Gene Transcripts

Gene Symbol
NOC3L
Entrez Gene ID
64318
Gene Name
NOC3 like DNA replication regulator
Gene Aliases
AD24, C10orf117, FAD24
Chromosome Location
Chr. 10: 94333226 - 94362976 on Build GRCh38
UniGene ID
Hs.74899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022451.10 NP_071896.8 2 286
GenBank mRNA AB077992.1 - 2 173
AK091246.1 - 2 243
AK292520.1 - 2 268
BC049850.1 - 2 236 5549517
BX505710.1 - 2 288

View Full Product Details