Gene Transcripts

Gene Symbol
IKZF4
Entrez Gene ID
64375
Gene Name
IKAROS family zinc finger 4
Gene Aliases
EOS, ZNFN1A4
Chromosome Location
Chr. 12: 56007647 - 56038435 on Build GRCh38
UniGene ID
Hs.553221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022465.3 NP_071910.3 6, 7 1462
XM_005269086.3 XP_005269143.1 8, 9 1004
XM_005269089.2 XP_005269146.1 6, 7 1106
XM_005269090.3 XP_005269147.1 7, 8 910
XM_005269093.2 XP_005269150.1 5, 6 1381
XM_011538664.1 XP_011536966.1 8, 9 1007
XM_011538666.2 XP_011536968.1 7, 8 855
XM_011538669.2 XP_011536971.1 6, 7 805
XM_017019805.1 XP_016875294.1 10, 11 1252
XM_017019806.1 XP_016875295.1 10, 11 1495
XM_017019807.1 XP_016875296.1 10, 11 1492
XM_017019808.1 XP_016875297.1 9, 10 1177
XM_017019809.1 XP_016875298.1 10, 11 1249
XM_017019810.1 XP_016875299.1 10, 11 1347
XM_017019811.1 XP_016875300.1 9, 10 1174
XM_017019812.1 XP_016875301.1 8, 9 1250
XM_017019813.1 XP_016875302.1 8, 9 1247
XM_017019814.1 XP_016875303.1 7, 8 1156
XM_017019815.1 XP_016875304.1 7, 8 1153
XM_017019816.1 XP_016875305.1 7, 8 913
GenBank mRNA AB058685.1 - 4, 5 1006
AF230809.1 - 5, 6 714
AK122880.1 - 5, 6 1229
AK314543.1 - 5, 6 827
BX647761.1 - 7, 8 1032

View Full Product Details