Gene Transcripts

Gene Symbol
C12orf43
Entrez Gene ID
64897
Gene Name
chromosome 12 open reading frame 43
Gene Aliases
Chromosome Location
Chr. 12: 121002422 - 121016502 on Build GRCh38
UniGene ID
Hs.528671
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286191.1 NP_001273120.1 2, 3 317
NM_001286192.1 NP_001273121.1 2, 3 224
NM_001286195.1 NP_001273124.1 2, 3 224
NM_001286196.1 NP_001273125.1 2, 3 224
NM_001286198.1 NP_001273127.1 3, 4 481
NM_022895.2 NP_075046.1 2, 3 224
NR_104409.1 2, 3 224
XM_017019829.1 XP_016875318.1 2, 3 308
GenBank mRNA AK022510.1 - 2, 3 219
AK222630.1 - 2, 3 209
AK225162.1 - 2, 3 219
AK301566.1 - 2, 3 209
AK307971.1 - 2, 3 319
BC014661.1 - 2, 3 198 4860511
BP201132.1 - 2, 3 204
DA014862.1 - 2, 3 213
DC297725.1 - 2, 3 211

View Full Product Details