Gene Transcripts

Gene Symbol
RSRC2
Entrez Gene ID
65117
Gene Name
arginine and serine rich coiled-coil 2
Gene Aliases
Chromosome Location
Chr. 12: 122504643 - 122527013 on Build GRCh38
UniGene ID
Hs.432996
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023012.5 NP_075388.2 10 1356
NR_036434.1 11 1437
NR_036435.1 11 1456
NR_036436.1 10 1378
XM_005253601.1 XP_005253658.1 10 1359
XM_005253602.2 XP_005253659.1 10 1330
XM_005253604.1 XP_005253661.1 11 1441
XM_011538688.1 XP_011536990.1 9 1269
XM_011538689.1 XP_011536991.1 13 1614
XM_017019830.1 XP_016875319.1 9 1266
XM_017019831.1 XP_016875320.1 11 1436
XM_017019832.1 XP_016875321.1 13 1592
XM_017019833.1 XP_016875322.1 11 1383
GenBank mRNA AB212664.1 - 10 1342
AK001883.1 - 10 1316
AK023985.1 - 7 946
AK056114.1 - 11 1378
BC008684.1 - 10 1266 3853928
BC067773.1 - 10 1326 5301415
BX640711.1 - 10 1302
BX647884.1 - 10 1345

View Full Product Details