Gene Transcripts
- Gene Symbol
- C1orf54
- Entrez Gene ID
- 79630
- Gene Name
- chromosome 1 open reading frame 54
- Gene Aliases
- Chromosome Location
- Chr. 1: 150272284 - 150280927 on Build GRCh38
- UniGene ID
- Hs.91283
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301039.1 |
NP_001287968.1
|
4, 5 |
342 |
|
NM_001301040.1 |
NP_001287969.1
|
4, 5 |
342 |
|
NM_001301041.1 |
NP_001287970.1
|
2, 3 |
128 |
|
NM_001301042.1 |
NP_001287971.1
|
3, 4 |
212 |
|
NM_024579.3 |
NP_078855.2
|
3, 4 |
231 |
|
XM_011509984.2 |
XP_011508286.1
|
3, 4 |
258 |
|
GenBank mRNA |
AA095731.1 |
- |
3, 4 |
199 |
|
AK026874.1 |
- |
3, 4 |
216 |
|
AX770997.1 |
- |
3, 4 |
229 |
|
AX770998.1 |
- |
4, 5 |
256 |
|
AX770999.1 |
- |
3, 4 |
231 |
|
BC017761.1 |
- |
3, 4 |
216 |
4617936 |
DA358536.1 |
- |
2, 3 |
139 |
|
View Full Product Details