Gene Transcripts
- Gene Symbol
- NOX5
- Entrez Gene ID
- 79400
- Gene Name
- NADPH oxidase, EF-hand calcium binding domain 5
- Gene Aliases
- Chromosome Location
- Chr. 15: 68930500 - 69057161 on Build GRCh38
- UniGene ID
- Hs.657932
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001184779.1 |
NP_001171708.1
|
13, 14 |
1864 |
|
NM_001184780.1 |
NP_001171709.1
|
13, 14 |
1991 |
|
NM_024505.3 |
NP_078781.3
|
13, 14 |
1948 |
|
NR_033671.2 |
|
14, 15 |
2179 |
|
NR_033672.1 |
|
14, 15 |
2095 |
|
GenBank mRNA |
AF317889.1 |
- |
10, 11 |
1379 |
|
AF325189.1 |
- |
14, 15 |
2070 |
|
AF325190.1 |
- |
14, 15 |
2154 |
|
AF353088.1 |
- |
13, 14 |
1843 |
|
AF353089.1 |
- |
13, 14 |
1927 |
|
AK026011.1 |
- |
10, 11 |
1433 |
|
AK074058.1 |
- |
11, 12 |
1617 |
|
AK074071.1 |
- |
12, 13 |
5066 |
|
AK314689.1 |
- |
13, 14 |
1927 |
|
AY166601.1 |
- |
9, 10 |
1174 |
|
BC125097.1 |
- |
13, 14 |
1857 |
40118286 |
BC125098.1 |
- |
14, 15 |
1961 |
40118288 |
DQ413001.1 |
- |
10, 11 |
1307 |
|
View Full Product Details