Gene Transcripts

Gene Symbol
LONRF3
Entrez Gene ID
79836
Gene Name
LON peptidase N-terminal domain and ring finger 3
Gene Aliases
RNF127
Chromosome Location
Chr. X: 118974614 - 119022925 on Build GRCh38
UniGene ID
Hs.144266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031855.2 NP_001027026.1 1 439
NM_024778.5 NP_079054.3 1 439
NR_110311.1 1 439
XM_005262476.2 XP_005262533.1 1 439
XM_011531398.2 XP_011529700.1 1 439
XM_017029845.1 XP_016885334.1 1 439
GenBank mRNA AK091777.1 - 1 435
AK290198.1 - 1 435
BC099847.1 - 1 303 40007729
BC100671.1 - 1 303 40007726
BC103491.1 - 1 303 40007723
DA249558.1 - 1 439

View Full Product Details