Gene Transcripts
- Gene Symbol
- LONRF3
- Entrez Gene ID
- 79836
- Gene Name
- LON peptidase N-terminal domain and ring finger 3
- Gene Aliases
- RNF127
- Chromosome Location
- Chr. X: 118974614 - 119022925 on Build GRCh38
- UniGene ID
- Hs.144266
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031855.2 |
NP_001027026.1
|
1, 2 |
979 |
|
NM_024778.5 |
NP_079054.3
|
1, 2 |
979 |
|
NR_110311.1 |
|
1, 2 |
979 |
|
XM_005262476.2 |
XP_005262533.1
|
1, 2 |
979 |
|
XM_011531398.2 |
XP_011529700.1
|
1, 2 |
979 |
|
XM_017029845.1 |
XP_016885334.1
|
1, 2 |
979 |
|
GenBank mRNA |
AK026265.1 |
- |
1, 2 |
231 |
|
AK091777.1 |
- |
1, 2 |
975 |
|
AK290198.1 |
- |
1, 2 |
975 |
|
BC099847.1 |
- |
1, 2 |
843 |
40007729 |
BC100671.1 |
- |
1, 2 |
843 |
40007726 |
BC103491.1 |
- |
1, 2 |
843 |
40007723 |
View Full Product Details