Gene Transcripts

Gene Symbol
NHEJ1
Entrez Gene ID
79840
Gene Name
non-homologous end joining factor 1
Gene Aliases
XLF
Chromosome Location
Chr. 2: 219075324 - 219160865 on Build GRCh38
UniGene ID
Hs.225988
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024782.2 NP_079058.1 7 896
GenBank mRNA AJ972687.1 - 7 840
AK022672.1 - 7 832
BC008210.2 - 3 222
BC012732.2 - 7 817 4026495
BC030986.1 - 7 849 4076851
CR457291.1 - 6 750

View Full Product Details