Gene Transcripts

Gene Symbol
NOL10
Entrez Gene ID
79954
Gene Name
nucleolar protein 10
Gene Aliases
PQBP5
Chromosome Location
Chr. 2: 10570766 - 10689991 on Build GRCh38
UniGene ID
Hs.222494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001261392.1 NP_001248321.1 3 311
NM_001261394.1 NP_001248323.1 3 311
NM_024894.3 NP_079170.2 3 311
NR_048552.1 2 265
XM_011510398.2 XP_011508700.1 3 315
XM_011510399.2 XP_011508701.1 2 253
XM_011510400.2 XP_011508702.1 3 299
XM_011510402.2 XP_011508704.1 3 299
XM_017004990.1 XP_016860479.1 1 53
GenBank mRNA AK024000.1 - 3 289
AK024137.1 - 3 296
AK290680.1 - 3 275
AK297169.1 - 3 299
AK299617.1 - 2 253
BC005125.2 - 3 291 3841643
DC346574.1 - 3 311

View Full Product Details