Gene Transcripts

Gene Symbol
PHF2
Entrez Gene ID
5253
Gene Name
PHD finger protein 2
Gene Aliases
CENP-35, GRC5, JHDM1E
Chromosome Location
Chr. 9: 93576627 - 93679587 on Build GRCh38
UniGene ID
Hs.211441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005392.3 NP_005383.3 7 1049
XM_005252051.2 XP_005252108.1 7 1043
GenBank mRNA AB014562.2 - 6 942
AF043725.1 - 7 1256
AL834263.1 - 7 900
BC152437.1 - 6 942

View Full Product Details