Gene Transcripts
- Gene Symbol
- WDR26
- Entrez Gene ID
- 80232
- Gene Name
- WD repeat domain 26
- Gene Aliases
- CDW2, GID7, MIP2
- Chromosome Location
- Chr. 1: 224385143 - 224434299 on Build GRCh38
- UniGene ID
- Hs.497873
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001115113.2 |
NP_001108585.2
|
6, 7 |
1176 |
|
NM_025160.6 |
NP_079436.4
|
6, 7 |
1224 |
|
XM_005273273.4 |
XP_005273330.1
|
6, 7 |
1125 |
|
XM_011544281.2 |
XP_011542583.1
|
6, 7 |
965 |
|
XM_011544282.2 |
XP_011542584.1
|
3, 4 |
287 |
|
XM_017002396.1 |
XP_016857885.1
|
6, 7 |
813 |
|
GenBank mRNA |
AB209887.1 |
- |
6, 7 |
1224 |
|
AK024669.1 |
- |
2, 3 |
238 |
|
AK225061.1 |
- |
2, 3 |
238 |
|
AK299230.1 |
- |
3, 4 |
409 |
|
AY221751.1 |
- |
6, 7 |
1014 |
|
AY304473.1 |
- |
6, 7 |
781 |
|
BC031471.2 |
- |
1, 2 |
167 |
|
BC034498.1 |
- |
7, 8 |
1110 |
|
BC052301.1 |
- |
5, 6 |
620 |
6420398 |
BC063817.1 |
- |
2, 3 |
286 |
|
EF011612.1 |
- |
5, 6 |
619 |
|
View Full Product Details