Gene Transcripts
- Gene Symbol
- LAS1L
- Entrez Gene ID
- 81887
- Gene Name
- LAS1 like, ribosome biogenesis factor
- Gene Aliases
- Las1-like, dJ475B7.2
- Chromosome Location
- Chr. X: 65512582 - 65534806 on Build GRCh38
- UniGene ID
- Hs.522675
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170649.1 |
NP_001164120.1
|
2, 3 |
461 |
|
NM_001170650.1 |
NP_001164121.1
|
1, 2 |
335 |
|
NM_031206.4 |
NP_112483.1
|
2, 3 |
461 |
|
XM_005262301.1 |
XP_005262358.1
|
2, 3 |
446 |
|
XM_005262304.1 |
XP_005262361.1
|
2, 3 |
446 |
|
XM_005262305.3 |
XP_005262362.1
|
2, 3 |
446 |
|
XM_005262306.3 |
XP_005262363.1
|
2, 3 |
446 |
|
XM_011531045.1 |
XP_011529347.1
|
1, 2 |
310 |
|
XM_011531046.1 |
XP_011529348.1
|
2, 3 |
446 |
|
XM_017029877.1 |
XP_016885366.1
|
2, 3 |
446 |
|
XM_017029878.1 |
XP_016885367.1
|
2, 3 |
446 |
|
XM_017029879.1 |
XP_016885368.1
|
2, 3 |
446 |
|
XM_017029880.1 |
XP_016885369.1
|
2, 3 |
446 |
|
GenBank mRNA |
AF116730.1 |
- |
2, 3 |
399 |
|
AK022587.1 |
- |
2, 3 |
411 |
|
AK074087.1 |
- |
2, 3 |
430 |
|
AK097383.1 |
- |
1, 2 |
285 |
|
AK225059.1 |
- |
2, 3 |
394 |
|
AK295591.1 |
- |
2, 3 |
411 |
|
BC014545.1 |
- |
2, 3 |
445 |
4079376 |
BC018610.1 |
- |
2, 3 |
409 |
3140243 |
BC019302.1 |
- |
2, 3 |
406 |
2822893 |
DA619280.1 |
- |
2, 3 |
462 |
|
View Full Product Details