Gene Transcripts

Gene Symbol
AKR1E2
Entrez Gene ID
83592
Gene Name
aldo-keto reductase family 1 member E2
Gene Aliases
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR
Chromosome Location
Chr. 10: 4824554 - 4908421 on Build GRCh38
UniGene ID
Hs.657944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040177.2 NP_001035267.1 10 1631
NM_001271021.1 NP_001257950.1 8 1460
NM_001271025.1 NP_001257954.1 7 1337
NR_073125.1 10 1700
NR_073126.1 10 1614
NR_073127.1 8 1529
XM_011519715.2 XP_011518017.1 10 1928
XM_011519718.2 XP_011518020.1 9 1845
XM_011519722.2 XP_011518024.1 9 1828
XM_011519724.2 XP_011518026.1 8 1756
XM_017016745.1 XP_016872234.1 9 1631
GenBank mRNA AF263242.1 - 10 1503
AK074211.1 - 10 1614
BC002862.2 - 10 1643 3941289

View Full Product Details