Gene Transcripts

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
Chromosome Location
Chr. 5: 93615130 - 94111699 on Build GRCh38
UniGene ID
Hs.600086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032042.5 NP_114431.2 2 281
NR_028080.1 2 281
XM_005272103.3 XP_005272160.1 2 247
XM_005272105.3 XP_005272162.1 2 246
XM_005272106.3 XP_005272163.1 2 247
XM_005272108.3 XP_005272165.1 2 276
XM_005272109.4 XP_005272166.1 2 247
XM_005272111.4 XP_005272168.1 2 247
XM_006714717.3 XP_006714780.1 2 273
XM_011543668.2 XP_011541970.1 2 247
XM_011543670.2 XP_011541972.1 2 247
XM_011543671.2 XP_011541973.1 2 247
XM_011543672.2 XP_011541974.1 1 52
XM_011543673.2 XP_011541975.1 1 52
XM_011543674.2 XP_011541976.1 2 247
XM_017009952.1 XP_016865441.1 1 52
XM_017009954.1 XP_016865443.1 2 247
XM_017009961.1 XP_016865450.1 2 191
GenBank mRNA AK295127.1 - 2 281
AK297471.1 - 1 43
AK312930.1 - 2 186
AL136630.1 - 2 120
BC020584.2 - 1 83 4398813
DC324758.1 - 2 281
DC374283.1 - 2 266

View Full Product Details