Gene Transcripts

Gene Symbol
QRICH2
Entrez Gene ID
84074
Gene Name
glutamine rich 2
Gene Aliases
Chromosome Location
Chr. 17: 76271296 - 76308690 on Build GRCh38
UniGene ID
Hs.252739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032134.2 NP_115510.1 4 432
XM_005257728.3 XP_005257785.1 4 1442
XM_011525344.2 XP_011523646.1 2 115
XM_017025206.1 XP_016880695.1 4 1442
XM_017025207.1 XP_016880696.1 3 1382
GenBank mRNA AL136774.1 - 4 432

View Full Product Details