Gene Transcripts

Gene Symbol
RNF26
Entrez Gene ID
79102
Gene Name
ring finger protein 26
Gene Aliases
Chromosome Location
Chr. 11: 119334500 - 119337314 on Build GRCh38
UniGene ID
Hs.524084
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032015.4 NP_114404.1 1 2065
GenBank mRNA AK075379.1 - 1 1823
AK122642.1 - 1 2038
BC000058.2 - 1 1834 3507662
BC007534.1 - 1 1805 3028875
BX647424.1 - 1 2038

View Full Product Details