Gene Transcripts

Gene Symbol
MND1
Entrez Gene ID
84057
Gene Name
meiotic nuclear divisions 1
Gene Aliases
GAJ
Chromosome Location
Chr. 4: 153344649 - 153415097 on Build GRCh38
UniGene ID
Hs.294088
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253861.1 NP_001240790.1 4 288
NM_032117.3 NP_115493.1 4 288
NR_045605.1 4 288
XM_005263275.2 XP_005263332.1 4 247
XM_017008688.1 XP_016864177.1 4 645
GenBank mRNA AK312172.1 - 4 247
AY028916.1 - 4 227
BC032142.2 - 4 242 5095005
BF975093.1 - 4 257
BG613292.1 - 4 250
BG758047.1 - 4 257
BM820228.1 - 4 288
BM833450.1 - 4 288

View Full Product Details