Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001201536.1 | NP_001188465.1 | 2 | 296 | |
NM_005681.3 | NP_005672.1 | 2 | 296 | ||
NM_139352.2 | NP_647603.1 | 2 | 296 | ||
XM_005273343.3 | XP_005273400.1 | 2 | 459 | ||
XM_006711612.1 | XP_006711675.1 | 2 | 459 | ||
XM_006711613.3 | XP_006711676.1 | 2 | 291 | ||
XM_017002758.1 | XP_016858247.1 | 2 | 459 | ||
XM_017002759.1 | XP_016858248.1 | 2 | 247 | ||
XM_017002760.1 | XP_016858249.1 | 2 | 243 | ||
GenBank mRNA | AK001054.1 | - | 2 | 148 | |
AK290970.1 | - | 2 | 241 | ||
AK299536.1 | - | 2 | 296 | ||
AK315740.1 | - | 2 | 280 | ||
BC013808.1 | - | 2 | 276 | 3918866 | |
L39060.1 | - | 1 | 111 |