Gene Transcripts

Gene Symbol
RELB
Entrez Gene ID
5971
Gene Name
RELB proto-oncogene, NF-kB subunit
Gene Aliases
I-REL, IREL, REL-B
Chromosome Location
Chr. 19: 45001449 - 45038198 on Build GRCh38
UniGene ID
Hs.654402
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006509.3 NP_006500.2 2, 3 285
XM_005259128.2 XP_005259185.1 2, 3 285
GenBank mRNA AK290594.1 - 2, 3 192
AL583128.3 - 2, 3 271
BC028013.1 - 2, 3 237 5215944
BI906507.1 - 2, 3 307
DA940587.1 - 2, 3 285
M83221.1 - 2, 3 298

View Full Product Details