Gene Transcripts

Gene Symbol
ZCCHC7
Entrez Gene ID
84186
Gene Name
zinc finger CCHC-type containing 7
Gene Aliases
AIR1, HSPC086
Chromosome Location
Chr. 9: 37120169 - 37358149 on Build GRCh38
UniGene ID
Hs.654700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289119.1 NP_001276048.1 2 498
NM_001289120.1 NP_001276049.1 2 521
NM_001289121.1 NP_001276050.1 2 639
NM_032226.2 NP_115602.2 2 494
XM_005251608.4 XP_005251665.1 2 1403
XM_005251612.2 XP_005251669.1 2 461
XM_011518050.1 XP_011516352.1 2 489
XM_017015197.1 XP_016870686.1 2 616
XM_017015198.1 XP_016870687.1 2 1102
XM_017015199.1 XP_016870688.1 2 463
XM_017015200.1 XP_016870689.1 2 736
GenBank mRNA AK026264.1 - 2 384
AK315128.1 - 2 511
BC005990.1 - 2 427
BC022434.1 - 3 587
BC036940.1 - 2 612 5399765
BP216150.1 - 2 521

View Full Product Details