Gene Transcripts

Gene Symbol
RETNLB
Entrez Gene ID
84666
Gene Name
resistin like beta
Gene Aliases
FIZZ1, FIZZ2, HXCP2, RELM-beta, RELMb, RELMbeta, XCP2
Chromosome Location
Chr. 3: 108755639 - 108757283 on Build GRCh38
UniGene ID
Hs.307047
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032579.2 NP_115968.1 1 214
GenBank mRNA AF290873.1 - 1 216
AF323084.1 - 1 223
AM050721.1 - 1 118
AY358655.1 - 1 215
BC069318.1 - 1 187 7262095
BC113502.1 - 1 148 8322554
BC113528.1 - 1 148 8322580

View Full Product Details