Gene Transcripts

Gene Symbol
MYO18B
Entrez Gene ID
84700
Gene Name
myosin XVIIIB
Gene Aliases
KFS4
Chromosome Location
Chr. 22: 25742133 - 26063847 on Build GRCh38
UniGene ID
Hs.417959
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318245.1 NP_001305174.1 3 417
NM_032608.6 NP_115997.5 3 417
XM_011530458.2 XP_011528760.1 3 427
XM_011530459.2 XP_011528761.1 3 398
XM_011530460.2 XP_011528762.1 2 271
XM_011530461.2 XP_011528763.1 3 427
XM_011530464.2 XP_011528766.1 3 427
XM_011530465.2 XP_011528767.1 2 184
XM_011530466.2 XP_011528768.1 3 428
XM_017029012.1 XP_016884501.1 3 427
XM_017029013.1 XP_016884502.1 3 475
XM_017029014.1 XP_016884503.1 3 427
XM_017029015.1 XP_016884504.1 3 427
XM_017029017.1 XP_016884506.1 3 426
GenBank mRNA AB075376.1 - 3 417
AJ310931.2 - 3 408
AY077700.1 - 3 327
BC144596.1 - 2 217
BC144597.1 - 2 217 9053127
BC150626.1 - 2 217 9056996

View Full Product Details