Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052874.4 | NP_443106.1 | 3 | 406 | |
XM_017022893.1 | XP_016878382.1 | 3 | 161 | ||
GenBank mRNA | AY028792.1 | - | 3 | 222 | |
AY995211.1 | - | 3 | 164 | ||
BC062298.1 | - | 3 | 218 | 6375788 | |
DN990202.1 | - | 3 | 400 |