Gene Transcripts
- Gene Symbol
- UPF3B
- Entrez Gene ID
- 65109
- Gene Name
- UPF3 regulator of nonsense transcripts homolog B (yeast)
- Gene Aliases
- HUPF3B, MRX62, MRXS14, RENT3B, UPF3BP1, UPF3BP2, UPF3BP3, UPF3X, Upf3p-X
- Chromosome Location
- Chr. X: 119805311 - 119853028 on Build GRCh38
- UniGene ID
- Hs.103832
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023010.3 |
NP_075386.1
|
1, 2 |
264 |
|
NM_080632.2 |
NP_542199.1
|
1, 2 |
264 |
|
XM_017029737.1 |
XP_016885226.1
|
1, 2 |
264 |
|
XM_017029738.1 |
XP_016885227.1
|
1, 2 |
264 |
|
XM_017029739.1 |
XP_016885228.1
|
1, 2 |
264 |
|
XM_017029740.1 |
XP_016885229.1
|
1, 2 |
264 |
|
GenBank mRNA |
AF318576.1 |
- |
1, 2 |
184 |
|
AK093728.1 |
- |
1, 2 |
202 |
|
AY013251.1 |
- |
1, 2 |
234 |
|
BC121017.1 |
- |
1, 2 |
207 |
40117102 |
BC121018.1 |
- |
1, 2 |
207 |
|
BI549935.1 |
- |
1, 2 |
244 |
|
DA949746.1 |
- |
1, 2 |
264 |
|
View Full Product Details