Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080675.3 | NP_542406.2 | 6, 7 | 491 | |
XM_011528573.1 | XP_011526875.1 | 7, 8 | 575 | ||
XM_011528574.1 | XP_011526876.1 | 5, 6 | 431 | ||
XM_011528575.1 | XP_011526877.1 | 3, 4 | 210 | ||
XM_011528576.1 | XP_011526878.1 | 7, 8 | 576 | ||
GenBank mRNA | AF401350.1 | - | 6, 7 | 491 | |
AK097733.1 | - | 4, 5 | 239 | ||
BC026118.2 | - | 6, 7 | 491 | 4824412 | |
BC029528.1 | - | 6, 7 | 484 | 5271065 |