Gene Transcripts

Gene Symbol
XAGE5
Entrez Gene ID
170627
Gene Name
X antigen family member 5
Gene Aliases
CT12.5, GAGED5, XAGE-5
Chromosome Location
Chr. X: 52811295 - 52818301 on Build GRCh38
UniGene ID
Hs.356874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130775.2 NP_570131.1 3 196
XM_017029320.1 XP_016884809.1 4 429
GenBank mRNA BC069129.1 - 3 197
BC103962.1 - 2 494 40014176

View Full Product Details