Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138697.3 | NP_619642.2 | 1, 2 | 295 | |
NM_177540.2 | NP_803884.1 | 1, 2 | 295 | ||
XM_011542206.2 | XP_011540508.1 | 1 | 847 | ||
XM_017002402.1 | XP_016857891.1 | 1 | 847 | ||
XM_017002403.1 | XP_016857892.1 | 1 | 847 | ||
GenBank mRNA | AK292014.1 | - | 1, 2 | 264 | |
AK293499.1 | - | 1, 2 | 164 | ||
AK315805.1 | - | 1, 2 | 264 | ||
BC136515.1 | - | 1, 2 | 295 | 9020505 | |
BC136516.1 | - | 1, 2 | 294 | 9020506 |