Gene Transcripts

Gene Symbol
ALG14
Entrez Gene ID
199857
Gene Name
ALG14, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CMS15
Chromosome Location
Chr. 1: 94981565 - 95072974 on Build GRCh38
UniGene ID
Hs.408927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305242.1 NP_001292171.1 5 537
NM_144988.3 NP_659425.1 4 500
NR_131032.1 3 348
XM_011540897.2 XP_011539199.1 5 607
GenBank mRNA AK289395.1 - 4 500
AW242346.1 - 5 451
BC011706.2 - 4 476 3689162
HY073066.1 - 3 312

View Full Product Details