Gene Transcripts

Gene Symbol
FBXO22
Entrez Gene ID
26263
Gene Name
F-box protein 22
Gene Aliases
FBX22, FISTC1
Chromosome Location
Chr. 15: 75903859 - 75937513 on Build GRCh38
UniGene ID
Hs.591115
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012170.3 NP_036302.1 2, 3 383
NM_147188.2 NP_671717.1 2, 3 383
GenBank mRNA AK024048.1 - 2, 3 364
AK223500.1 - 2, 3 340
AY005144.1 - 2, 3 278
BC020204.1 - 2, 3 383 4546640
BC032540.1 - 2, 3 301 5529506
BC041691.1 - 2, 3 350 5246992
BG328756.1 - 2, 3 383

View Full Product Details