Gene Transcripts
- Gene Symbol
- FRMD6
- Entrez Gene ID
- 122786
- Gene Name
- FERM domain containing 6
- Gene Aliases
- C14orf31, EX1, Willin, c14_5320
- Chromosome Location
- Chr. 14: 51489100 - 51730727 on Build GRCh38
- UniGene ID
- Hs.434914
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042481.2 |
NP_001035946.1
|
5, 6 |
787 |
|
NM_152330.3 |
NP_689543.1
|
4, 5 |
563 |
|
XM_006720030.1 |
XP_006720093.1
|
4, 5 |
660 |
|
XM_017020968.1 |
XP_016876457.1
|
4, 5 |
513 |
|
GenBank mRNA |
AK126521.1 |
- |
2, 3 |
208 |
|
AK315764.1 |
- |
4, 5 |
484 |
|
AL833158.1 |
- |
5, 6 |
773 |
|
BC020521.2 |
- |
4, 5 |
511 |
3914276 |
BX161430.1 |
- |
4, 5 |
563 |
|
BX647863.1 |
- |
4, 5 |
475 |
|
View Full Product Details