Gene Transcripts

Gene Symbol
MAB21L3
Entrez Gene ID
126868
Gene Name
mab-21 like 3
Gene Aliases
C1orf161
Chromosome Location
Chr. 1: 116072744 - 116135240 on Build GRCh38
UniGene ID
Hs.376194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152367.2 NP_689580.2 3 360
XM_011540623.2 XP_011538925.1 3 492
XM_011540625.2 XP_011538927.1 4 588
XM_011540626.2 XP_011538928.1 3 414
XM_017000260.1 XP_016855749.1 4 1825
GenBank mRNA AK096035.1 - 3 360
BC128148.1 - 2 254 40112561
BC128149.1 - 2 254 40112562

View Full Product Details