Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001160011.1 | NP_001153483.1 | 3 | 450 | |
NM_152996.2 | NP_694541.2 | 3 | 450 | ||
XM_006710553.3 | XP_006710616.1 | 4 | 446 | ||
XM_017000937.1 | XP_016856426.1 | 3 | 3592 | ||
XM_017000938.1 | XP_016856427.1 | 3 | 3604 | ||
XM_017000939.1 | XP_016856428.1 | 3 | 3602 | ||
XM_017000940.1 | XP_016856429.1 | 4 | 709 | ||
XM_017000941.1 | XP_016856430.1 | 3 | 1364 | ||
XM_017000942.1 | XP_016856431.1 | 3 | 1169 | ||
GenBank mRNA | AJ507291.1 | - | 3 | 308 | |
AK091215.1 | - | 3 | 435 | ||
AK297363.1 | - | 2 | 219 | ||
AY358540.1 | - | 3 | 409 | ||
BC039520.1 | - | 2 | 161 | ||
BC059363.1 | - | 3 | 421 | 30341233 |