Gene Transcripts

Gene Symbol
FPR2
Entrez Gene ID
2358
Gene Name
formyl peptide receptor 2
Gene Aliases
ALXR, FMLP-R-II, FMLPX, FPR2A, FPRH1, FPRH2, FPRL1, HM63, LXA4R
Chromosome Location
Chr. 19: 51752026 - 51770526 on Build GRCh38
UniGene ID
Hs.99855
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005738.1 NP_001005738.1 2 657
NM_001462.3 NP_001453.1 2 755
XM_006723120.3 XP_006723183.1 5 1488
XM_017026542.1 XP_016882031.1 4 1332
GenBank mRNA AF054013.1 - 1 612
AK290557.1 - 3 803
BC029125.2 - 1 626 5180065
BC071722.1 - 2 927 5212947
D10922.1 - 2 646
M76672.1 - 1 612
M84562.1 - 2 1384
M88107.1 - 2 651
U81501.1 - 1 632
X63819.1 - 2 765

View Full Product Details