Gene Transcripts

Gene Symbol
ADIG
Entrez Gene ID
149685
Gene Name
adipogenin
Gene Aliases
SMAF1
Chromosome Location
Chr. 20: 38581195 - 38588463 on Build GRCh38
UniGene ID
Hs.368028
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018082.2 NP_001018092.1 2 396
GenBank mRNA BC029594.1 - 2 396
BC119704.2 - 2 390 40117136
BC119705.2 - 2 390 40117137
BX105371.1 - 2 236

View Full Product Details