Gene Transcripts

Gene Symbol
RPS28
Entrez Gene ID
6234
Gene Name
ribosomal protein S28
Gene Aliases
DBA15, S28
Chromosome Location
Chr. 19: 8321500 - 8322396 on Build GRCh38
UniGene ID
Hs.153177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031.4 NP_001022.1 1 353
GenBank mRNA AK293636.1 - 3 602
AK301638.1 - 2 782
AU126783.1 - 4 353
BC000354.1 - 1 339 2964364
BC018810.1 - 1 339
BC021239.2 - 1 332 4309752
BC070217.1 - 1 355 4248660
BC070218.1 - 1 350 6649143
L05091.1 - 4 337
U58682.1 - 4 334

View Full Product Details