Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001018.4 | NP_001009.1 | 3, 4 | 388 | |
NM_001308226.1 | NP_001295155.1 | 3, 4 | 479 | ||
GenBank mRNA | BC064908.1 | - | 1 | 332 | |
BC105810.1 | - | 3, 4 | 366 | 6744553 | |
BC141832.1 | - | 1 | 332 | 40148744 | |
BE798984.1 | - | 3, 4 | 568 | ||
BG253776.1 | - | 1 | 327 | ||
BG705752.1 | - | 3, 4 | 486 | ||
BM558707.1 | - | 3, 4 | 467 | ||
BU602036.1 | - | 3, 4 | 389 | ||
J02984.1 | - | 3, 4 | 355 |