Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006161.2 | NP_006152.2 | 1 | 940 | |
GenBank mRNA | AB593107.1 | - | 1 | 952 | |
AK314280.1 | - | 1 | 952 | ||
BC008687.1 | - | 1 | 940 | 3872172 | |
BC028226.1 | - | 1 | 940 | 5241507 | |
BT019366.1 | - | 1 | 681 |