Gene Transcripts

Gene Symbol
TLX2
Entrez Gene ID
3196
Gene Name
T-cell leukemia homeobox 2
Gene Aliases
HOX11L1, NCX
Chromosome Location
Chr. 2: 74514469 - 74517148 on Build GRCh38
UniGene ID
Hs.168586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016170.4 NP_057254.1 3 1156
GenBank mRNA AB008501.1 - 3 818
AB593086.1 - 4 843
BC006356.2 - 3 1141 4125397
BC006396.1 - 2 1413
BC018611.1 - 4 752

View Full Product Details