Gene Transcripts

Gene Symbol
WWC3
Entrez Gene ID
55841
Gene Name
WWC family member 3
Gene Aliases
BM042
Chromosome Location
Chr. X: 10015755 - 10144478 on Build GRCh38
UniGene ID
Hs.527524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015691.3 NP_056506.2 6 668
GenBank mRNA AB033106.1 - 4 291
AK292961.1 - 5 550
AL353937.1 - 2 148
BC032447.1 - 3 203
KC987947.1 - 6 845

View Full Product Details