Gene Transcripts
- Gene Symbol
- ATCAY
- Entrez Gene ID
- 85300
- Gene Name
- ATCAY, caytaxin
- Gene Aliases
- BNIP-H, CLAC
- Chromosome Location
- Chr. 19: 3880620 - 3928082 on Build GRCh38
- UniGene ID
- Hs.418055
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033064.4 |
NP_149053.1
|
3, 4 |
565 |
|
GenBank mRNA |
AB058775.1 |
- |
3, 4 |
511 |
|
AK027889.1 |
- |
3, 4 |
500 |
|
AK092309.1 |
- |
3, 4 |
364 |
|
AK125457.1 |
- |
3, 4 |
208 |
|
AK299522.1 |
- |
4, 5 |
571 |
|
AY220297.1 |
- |
2, 3 |
135 |
|
BC026217.1 |
- |
3, 4 |
507 |
4153341 |
DA151785.1 |
- |
3, 4 |
544 |
|
KU178814.1 |
- |
2, 3 |
135 |
|
KU178815.1 |
- |
2, 3 |
135 |
|
View Full Product Details