Gene Transcripts

Gene Symbol
KLK15
Entrez Gene ID
55554
Gene Name
kallikrein related peptidase 15
Gene Aliases
ACO, HSRNASPH
Chromosome Location
Chr. 19: 50825289 - 50834337 on Build GRCh38
UniGene ID
Hs.567535
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277081.1 NP_001264010.1 5 785
NM_001277082.1 NP_001264011.1 4 648
NM_017509.3 NP_059979.2 5 788
NR_102274.1 5 667
XM_006723265.3 XP_006723328.1 6 1589
XM_011527085.2 XP_011525387.1 6 1585
XM_011527087.2 XP_011525389.1 5 1338
XM_011527089.2 XP_011525391.1 5 1459
GenBank mRNA AF303046.1 - 5 787
AY373373.1 - 4 877
AY373374.1 - 4 740
BC069480.1 - 5 785 7262190
BC069507.1 - 5 788 7262202
BC069518.1 - 5 785 7262214
BC126137.1 - 5 778 8991853
BC144045.1 - 5 657
BC144046.1 - 5 775 9052566
BC144047.1 - 4 638
X75363.1 - 1 1337

View Full Product Details