Gene Transcripts

Gene Symbol
MAGED4B
Entrez Gene ID
81557
Gene Name
MAGE family member D4B
Gene Aliases
Chromosome Location
Chr. X: 52061827 - 52069272 on Build GRCh38
UniGene ID
Hs.522650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242362.1 NP_001229291.1 6 1526
NM_030801.3 NP_110428.2 6 1505
NM_177535.2 NP_803879.1 6 1475
NM_177537.2 NP_803881.1 6 1475
XM_006724607.3 XP_006724670.1 6 1491
XM_006724608.3 XP_006724671.1 6 1517
XM_011530822.1 XP_011529124.1 6 1432
XM_011530823.2 XP_011529125.1 6 1432
XM_017029876.1 XP_016885365.1 6 1491
GenBank mRNA AB040527.1 - 6 1386
AB040528.1 - 6 1386
AB040529.1 - 5 1887
AB058762.2 - 6 1481
AF329733.1 - 5 1154
AK094541.1 - 3 296
AK098830.1 - 7 1388
AK289458.1 - 6 1440
AL136582.1 - 6 1424
BC001207.1 - 6 1473
BC063838.1 - 6 1387 6185437
BC081566.1 - 6 1399 6292139

Gene Symbol
MAGED4
Entrez Gene ID
728239
Gene Name
MAGE family member D4
Gene Aliases
MAGE-E1, MAGE1, MAGEE1
Chromosome Location
Chr. X: 52184823 - 52192268 on Build GRCh38
UniGene ID
Hs.522650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098800.2 NP_001092270.1 6 1475
NM_001272061.1 NP_001258990.1 6 1526
NM_001272062.1 NP_001258991.1 6 1475
NM_001272063.1 NP_001258992.1 6 1505

View Full Product Details