Gene Transcripts

Gene Symbol
TOMM6
Entrez Gene ID
100188893
Gene Name
translocase of outer mitochondrial membrane 6
Gene Aliases
OBTP, TOM6
Chromosome Location
Chr. 6: 41787443 - 41789896 on Build GRCh38
UniGene ID
Hs.731633
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134493.1 NP_001127965.1 2 451
GenBank mRNA AB435319.1 - 2 196
AF216754.1 - 2 222
AJ420506.1 - 2 1160
BC015975.1 - 2 233
BC018624.1 - 2 215
BC022274.1 - 2 209
BC107435.1 - 2 227
BM783950.1 - 2 561
L34839.1 - 2 173

View Full Product Details