Gene Transcripts

Gene Symbol
C14orf80
Entrez Gene ID
283643
Gene Name
chromosome 14 open reading frame 80
Gene Aliases
Chromosome Location
Chr. 14: 105489855 - 105499248 on Build GRCh38
UniGene ID
Hs.720306
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134875.1 NP_001128347.1 5 758
GenBank mRNA BC016028.1 - 4 426
BX248074.1 - 5 679

View Full Product Details