Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001074.2 | NP_001065.2 | 1 | 309 | |
XM_011532229.2 | XP_011530531.1 | 1 | 310 | ||
XM_011532230.1 | XP_011530532.1 | 1 | 310 | ||
GenBank mRNA | AK223142.1 | - | 1 | 283 | |
AK313190.1 | - | 1 | 310 | ||
BC030974.1 | - | 1 | 309 | 4593426 | |
J05428.1 | - | 1 | 277 |