Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002443.3 | NP_002434.1 | 4 | 284 | |
NM_138634.2 | NP_619540.1 | 3 | 178 | ||
GenBank mRNA | AK312108.1 | - | 4 | 284 | |
BC005257.1 | - | 4 | 284 | 3950847 | |
BF680662.1 | - | 3 | 179 | ||
BM856408.1 | - | 4 | 295 | ||
BT006816.1 | - | 3 | 252 | ||
CR457015.1 | - | 3 | 252 | ||
M15885.1 | - | 3 | 263 | ||
S67815.1 | - | 4 | 272 | ||
U22178.1 | - | 2 | 157 | ||
U78976.1 | - | 3 | 276 |