Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002909.4 | NP_002900.2 | 4 | 381 | |
GenBank mRNA | AF172331.1 | - | 3 | 328 | |
AK291981.1 | - | 4 | 349 | ||
AK307292.1 | - | 3 | 785 | ||
BC005350.2 | - | 3 | 325 | 3950349 | |
BM769434.1 | - | 4 | 393 | ||
M18963.1 | - | 3 | 333 | ||
M27190.1 | - | 3 | 360 |