Gene Transcripts

Gene Symbol
TLR3
Entrez Gene ID
7098
Gene Name
toll like receptor 3
Gene Aliases
CD283, IIAE2
Chromosome Location
Chr. 4: 186069155 - 186086724 on Build GRCh38
UniGene ID
Hs.657724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003265.2 NP_003256.1 3 610
GenBank mRNA AB445631.1 - 2 509
AK314208.1 - 3 609
BC059372.1 - 3 609
BC094737.1 - 3 609 30531515
BC096333.1 - 3 587 40008349
BC096334.3 - 3 587 40008350
BC096335.1 - 3 587 40008351
DA374150.1 - 3 610
DQ445682.1 - 3 580
U88879.1 - 3 582

View Full Product Details