Gene Transcripts

Gene Symbol
Pcyt1b
Entrez Gene ID
286936
Gene Name
phosphate cytidylyltransferase 1, choline, beta
Gene Aliases
CCT-beta, CTB, Cctbeta
Chromosome Location
Chr. X: 62727617 - 62818516 on Build Rnor_6.0
UniGene ID
Rn.48742
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173151.1 NP_775174.1 1 102
GenBank mRNA AF190256.1 - 1 102

View Full Product Details